missing translation for 'onlineSavingsMsg'
Learn More

4-Heptylbenzonitrile 98.0+%, TCI America™

Catalog No. H07785ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07785ML 5 mL
H07781ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07785ML Supplier TCI America Supplier No. H07785ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60484-67-5
Molecular Formula C14H19N
Molecular Weight (g/mol) 201.313
MDL Number MFCD00143348
InChI Key XTIKBCXMOYZUMG-UHFFFAOYSA-N
Synonym 1-(4-Cyanophenyl)heptane
PubChem CID 575863
IUPAC Name 4-heptylbenzonitrile
SMILES CCCCCCCC1=CC=C(C=C1)C#N

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 201.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Heptylbenzonitrile
EINECSNumber (3)-3541
TSCA No