missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Heptylaniline 98.0+%, TCI America™
$130.48 - $432.46
Chemical Identifiers
| CAS | 37529-27-4 |
|---|---|
| Molecular Formula | C13H21N |
| Molecular Weight (g/mol) | 191.32 |
| MDL Number | MFCD00007928 |
| InChI Key | BNEWZYZRLNNWNR-UHFFFAOYSA-N |
| Synonym | p-heptylaniline, 4-n-heptylaniline, benzenamine, 4-heptyl, 4-heptylphenylamine, acmc-209ito, 4-heptylaniline |
| PubChem CID | 142169 |
| IUPAC Name | 4-heptylaniline |
| SMILES | CCCCCCCC1=CC=C(N)C=C1 |
Chemical Identifiers
| 37529-27-4 | |
| 191.32 | |
| BNEWZYZRLNNWNR-UHFFFAOYSA-N | |
| 142169 | |
| CCCCCCCC1=CC=C(N)C=C1 |
| C13H21N | |
| MFCD00007928 | |
| p-heptylaniline, 4-n-heptylaniline, benzenamine, 4-heptyl, 4-heptylphenylamine, acmc-209ito, 4-heptylaniline | |
| 4-heptylaniline |
Specifications
| 37529-27-4 | |
| C13H21N | |
| 5 mL | |
| BNEWZYZRLNNWNR-UHFFFAOYSA-N | |
| 4-heptylaniline | |
| 142169 | |
| ≥98.0% (GC,T) | |
| 4-Heptylaniline |
| 286°C | |
| MFCD00007928 | |
| p-heptylaniline, 4-n-heptylaniline, benzenamine, 4-heptyl, 4-heptylphenylamine, acmc-209ito, 4-heptylaniline | |
| CCCCCCCC1=CC=C(N)C=C1 | |
| 191.32 | |
| 191.32 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0206
TSCA : No