missing translation for 'onlineSavingsMsg'
Learn More

4-Heptylaniline 98.0+%, TCI America™

Catalog No. H05885ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05885ML 5 mL
H058825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05885ML Supplier TCI America Supplier No. H05885ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37529-27-4
Molecular Formula C13H21N
Molecular Weight (g/mol) 191.32
MDL Number MFCD00007928
InChI Key BNEWZYZRLNNWNR-UHFFFAOYSA-N
Synonym p-heptylaniline, 4-n-heptylaniline, benzenamine, 4-heptyl, 4-heptylphenylamine, acmc-209ito, 4-heptylaniline
PubChem CID 142169
IUPAC Name 4-heptylaniline
SMILES CCCCCCCC1=CC=C(N)C=C1

Specifications

Boiling Point 286°C
Quantity 5 mL
Formula Weight 191.32
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Heptylaniline
EINECSNumber (3)-0206
TSCA No