missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Heptylacetophenone 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 163°C |
| Quantity | 25 mL |
| Formula Weight | 218.34 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4′-Heptylacetophenone |
Safety and Handling
| EINECSNumber | (3)-3540 |
| TSCA | Yes |