missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Heptanone 97.0+%, TCI America™
$41.02 - $391.44
Chemical Identifiers
| CAS | 123-19-3 |
|---|---|
| Molecular Formula | C7H14O |
| Molecular Weight (g/mol) | 114.188 |
| MDL Number | MFCD00009403 |
| InChI Key | HCFAJYNVAYBARA-UHFFFAOYSA-N |
| Synonym | 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon |
| PubChem CID | 31246 |
| IUPAC Name | heptan-4-one |
| SMILES | CCCC(=O)CCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H003925ML
|
TCI America
H003925ML |
25 mL |
Each for $41.02
|
|
|||||
|
H0039500ML
|
TCI America
H0039500ML |
500 mL |
Each for $391.44
|
|
|||||
Chemical Identifiers
| 123-19-3 | |
| 114.188 | |
| HCFAJYNVAYBARA-UHFFFAOYSA-N | |
| 31246 | |
| CCCC(=O)CCC |
| C7H14O | |
| MFCD00009403 | |
| 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon | |
| heptan-4-one |
Specifications
| 123-19-3 | |
| 143°C | |
| MFCD00009403 | |
| 2710 | |
| HCFAJYNVAYBARA-UHFFFAOYSA-N | |
| heptan-4-one | |
| 31246 | |
| ≥97.0% (GC) | |
| 4-Heptanone |
| Yellow | |
| C7H14O | |
| 25 mL | |
| 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon | |
| CCCC(=O)CCC | |
| 114.188 | |
| 114.19 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2475
RTECSNumber : MJ5600000
TSCA : Yes