missing translation for 'onlineSavingsMsg'
Learn More

4-Heptanone 97.0+%, TCI America™

Catalog No. H003925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H003925ML 25 mL
H0039500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H003925ML Supplier TCI America Supplier No. H003925ML

Chemical Identifiers

CAS 123-19-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00009403
InChI Key HCFAJYNVAYBARA-UHFFFAOYSA-N
Synonym 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon
PubChem CID 31246
IUPAC Name heptan-4-one
SMILES CCCC(=O)CCC

Specifications

Color Yellow
Boiling Point 143°C
Quantity 25 mL
UN Number 2710
Formula Weight 114.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Heptanone
EINECSNumber (2)-2475
RTECSNumber MJ5600000
TSCA Yes