Learn More
4-Heptanone, 98%
CAS: 123-19-3 | C7H14O | 114.188 g/mol
Supplier: Thermo Scientific Chemicals B21222AE
Description
4-Heptanone is used as a solvent for nitrocellulose as well as in organic synthesis. It is involved in the preparation of flavorings. It is used as an intermediate in the synthesis of active pharmaceutical ingredients. Further, it is used in paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Heptanone is used as a solvent for nitrocellulose as well as in organic synthesis. It is involved in the preparation of flavorings. It is used as an intermediate in the synthesis of active pharmaceutical ingredients. Further, it is used in paints and coatings.
Solubility
Miscible with alcohol, ligroin and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications
| 4-Heptanone | |
| -33°C | |
| 143°C to 144°C | |
| C7H14O | |
| MFCD00009403 | |
| UN2710 | |
| 14,3345 | |
| Miscible with alcohol,ligroin and ether. Immiscible with water. | |
| CCCC(=O)CCC | |
| 114.188 | |
| 114.19 |
| 123-19-3 | |
| 0.818 | |
| 48°C (118°F) | |
| 1.407 | |
| 100 mL | |
| 1699049 | |
| 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon | |
| HCFAJYNVAYBARA-UHFFFAOYSA-N | |
| heptan-4-one | |
| 31246 | |
| 98% |
Chemical Identifiers
| 123-19-3 | |
| 114.188 | |
| HCFAJYNVAYBARA-UHFFFAOYSA-N | |
| 31246 | |
| CCCC(=O)CCC |
| C7H14O | |
| MFCD00009403 | |
| 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon | |
| heptan-4-one |
Safety and Handling
GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIPROPYL KETONE
EINECSNumber : 204-608-9
RTECSNumber : MJ5600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only