missing translation for 'onlineSavingsMsg'
Learn More

4-Heptanone, 98%

Catalog No. AAB21222AU
Change view
Click to view available options
Quantity:
100 mL
250 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21222AU 1000 mL
AAB21222AE 100 mL
AAB21222AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB21222AU Supplier Thermo Scientific Chemicals Supplier No. B21222AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 123-19-3 | C7H14O | 114.188 g/mol

4-Heptanone is used as a solvent for nitrocellulose as well as in organic synthesis. It is involved in the preparation of flavorings. It is used as an intermediate in the synthesis of active pharmaceutical ingredients. Further, it is used in paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Heptanone is used as a solvent for nitrocellulose as well as in organic synthesis. It is involved in the preparation of flavorings. It is used as an intermediate in the synthesis of active pharmaceutical ingredients. Further, it is used in paints and coatings.

Solubility
Miscible with alcohol, ligroin and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.

Chemical Identifiers

CAS 123-19-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00009403
InChI Key HCFAJYNVAYBARA-UHFFFAOYSA-N
Synonym 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon
PubChem CID 31246
IUPAC Name heptan-4-one
SMILES CCCC(=O)CCC

Specifications

Melting Point -33°C
Density 0.818
Boiling Point 143°C to 144°C
Flash Point 48°C (118°F)
Refractive Index 1.407
Quantity 1000 mL
UN Number UN2710
Beilstein 1699049
Merck Index 14,3345
Solubility Information Miscible with alcohol,ligroin and ether. Immiscible with water.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 4-Heptanone
Show More Show Less
Hazard Category H226-H332
Hazard Statement GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIPROPYL KETONE
EINECSNumber 204-608-9
RTECSNumber MJ5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only