missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenyl beta-D-allopyranoside, 98%, Thermo Scientific Chemicals

$126.93 - $126.93

Chemical Identifiers

CAS 80154-34-3
Molecular Formula C13H16O7
Molecular Weight (g/mol) 284.264
MDL Number MFCD00210992
InChI Key OLZAGZCCJJBKNZ-SYLRKERUSA-N
Synonym helicide, helicid, hilicidum, 4-formylphenyl beta-d-allopyranoside, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde, 4-formylphenyl ss-d-allopyranoside, 4-beta-d-allopyranosyloxy benzaldehyde, 4-formylphenyl b-d-allopyranoside
PubChem CID 12896796
IUPAC Name 4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde
SMILES C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6610206
View Documents
Thermo Scientific Chemicals
J6610206
5 g
Each for $126.93
Only null left
 
Description

Description

It is used in the treatment of ischemic cardiovascular and cerebrovascular diseases. Also used in Chinese medicine in the treatment of headache and insomnia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in hot water. Slightly soluble in water, ethanol or carbinol. Insoluble in aether or chloroform.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

80154-34-3
284.264
OLZAGZCCJJBKNZ-SYLRKERUSA-N
12896796
C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O
C13H16O7
MFCD00210992
helicide, helicid, hilicidum, 4-formylphenyl beta-d-allopyranoside, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde, 4-formylphenyl ss-d-allopyranoside, 4-beta-d-allopyranosyloxy benzaldehyde, 4-formylphenyl b-d-allopyranoside
4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde

Specifications

80154-34-3
White
MFCD00210992
helicide, helicid, hilicidum, 4-formylphenyl beta-d-allopyranoside, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde, 4-formylphenyl ss-d-allopyranoside, 4-beta-d-allopyranosyloxy benzaldehyde, 4-formylphenyl b-d-allopyranoside
OLZAGZCCJJBKNZ-SYLRKERUSA-N
C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O
284.264
284.26
Powder
190°C
C13H16O7
5 g
Soluble in hot water. Slightly soluble in water,ethanol or carbinol. Insoluble in aether or chloroform.
−105° (c=0.8 in Water)
4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde
12896796
98%
4-Formylphenyl beta-D-allopyranoside
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only