missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenyl beta-D-allopyranoside, 98%, Thermo Scientific Chemicals

Catalog No. AAJ6610206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6610206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6610206 Supplier Thermo Scientific Chemicals Supplier No. J6610206

It is used in the treatment of ischemic cardiovascular and cerebrovascular diseases. Also used in Chinese medicine in the treatment of headache and insomnia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in hot water. Slightly soluble in water, ethanol or carbinol. Insoluble in aether or chloroform.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 80154-34-3
Molecular Formula C13H16O7
Molecular Weight (g/mol) 284.264
MDL Number MFCD00210992
InChI Key OLZAGZCCJJBKNZ-SYLRKERUSA-N
Synonym helicide, helicid, hilicidum, 4-formylphenyl beta-d-allopyranoside, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde, 4-formylphenyl ss-d-allopyranoside, 4-beta-d-allopyranosyloxy benzaldehyde, 4-formylphenyl b-d-allopyranoside
PubChem CID 12896796
IUPAC Name 4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde
SMILES C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 190°C
Color White
Quantity 5 g
Solubility Information Soluble in hot water. Slightly soluble in water,ethanol or carbinol. Insoluble in aether or chloroform.
Optical Rotation −105° (c=0.8 in Water)
Formula Weight 284.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Formylphenyl beta-D-allopyranoside
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only