missing translation for 'onlineSavingsMsg'
Learn More

4-Formylmorpholine 99.0+%, TCI America™

$42.36 - $167.49

Chemical Identifiers

CAS 4394-85-8
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00006170
InChI Key LCEDQNDDFOCWGG-UHFFFAOYSA-N
Synonym 4-formylmorpholine, n-formylmorpholine, 4-morpholinecarboxaldehyde, morpholine, 4-formyl, n-formylmorfolin, 4-morpholinecarbaldehyde, n-formylmorfolin czech, unii-d1e63xo4rh, n-formyhnorpholine, n-formyl morpholin
PubChem CID 20417
ChEBI CHEBI:43989
IUPAC Name morpholine-4-carbaldehyde
SMILES C1COCCN1C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F015725G
View Documents
TCI America
F015725G
25 g
Each for $42.36
Only null left
 
F0157500G
View Documents
TCI America
F0157500G
500 g
Each for $167.49
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4394-85-8
115.132
LCEDQNDDFOCWGG-UHFFFAOYSA-N
20417
morpholine-4-carbaldehyde
C5H9NO2
MFCD00006170
4-formylmorpholine, n-formylmorpholine, 4-morpholinecarboxaldehyde, morpholine, 4-formyl, n-formylmorfolin, 4-morpholinecarbaldehyde, n-formylmorfolin czech, unii-d1e63xo4rh, n-formyhnorpholine, n-formyl morpholin
CHEBI:43989
C1COCCN1C=O

Specifications

4394-85-8
237°C
MFCD00006170
4-formylmorpholine, n-formylmorpholine, 4-morpholinecarboxaldehyde, morpholine, 4-formyl, n-formylmorfolin, 4-morpholinecarbaldehyde, n-formylmorfolin czech, unii-d1e63xo4rh, n-formyhnorpholine, n-formyl morpholin
C1COCCN1C=O
115.132
CHEBI:43989
≥99.0% (GC)
4-Formylmorpholine
White
C5H9NO2
25 g
LCEDQNDDFOCWGG-UHFFFAOYSA-N
morpholine-4-carbaldehyde
20417
115.13
Crystalline Lumps
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6646

RTECSNumber : QD9694000

TSCA : Yes

SDS
Documents

Documents