missing translation for 'onlineSavingsMsg'
Learn More

4-Formylmorpholine 99.0+%, TCI America™

Catalog No. F0157500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0157500G 500 g
F015725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0157500G Supplier TCI America Supplier No. F0157500G

Chemical Identifiers

CAS 4394-85-8
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00006170
InChI Key LCEDQNDDFOCWGG-UHFFFAOYSA-N
Synonym 4-formylmorpholine, n-formylmorpholine, 4-morpholinecarboxaldehyde, morpholine, 4-formyl, n-formylmorfolin, 4-morpholinecarbaldehyde, n-formylmorfolin czech, unii-d1e63xo4rh, n-formyhnorpholine, n-formyl morpholin
PubChem CID 20417
ChEBI CHEBI:43989
IUPAC Name morpholine-4-carbaldehyde
SMILES C1COCCN1C=O

Specifications

Color White
Boiling Point 237°C
Quantity 500 g
Formula Weight 115.13
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Formylmorpholine
EINECSNumber (5)-6646
RTECSNumber QD9694000
TSCA Yes