missing translation for 'onlineSavingsMsg'
Learn More

4-Formylmorpholine, 99%

Catalog No. AAB2331522
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2331522 100 g
AAB2331536 500 g
AAB233150E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2331522 Supplier Thermo Scientific Chemicals Supplier No. B2331522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4394-85-8 | C5H9NO2 | 115.132 g/mol

4-Formylmorpholine is widely used as a solvent and extracting agent for aromatics. It is used as a scrubbing agent for acidic gases. It is employed in the preparation of adenine hydrochloride labeled with 14C, 4-(trifluoromethyl)morpholine, 3-alkyl-2-chloro-5,6-dihydrobenzaldehyde and terphenyl dialdehyde. It plays an important role in the desulfurization of natural gas, synthetic gas, flue gas and gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Formylmorpholine is widely used as a solvent and extracting agent for aromatics. It is used as a scrubbing agent for acidic gases. It is employed in the preparation of adenine hydrochloride labeled with 14C, 4-(trifluoromethyl)morpholine, 3-alkyl-2-chloro-5,6-dihydrobenzaldehyde and terphenyl dialdehyde. It plays an important role in the desulfurization of natural gas, synthetic gas, flue gas and gasoline.

Solubility
Miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4394-85-8
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00006170
InChI Key LCEDQNDDFOCWGG-UHFFFAOYSA-N
Synonym 4-formylmorpholine, n-formylmorpholine, 4-morpholinecarboxaldehyde, morpholine, 4-formyl, n-formylmorfolin, 4-morpholinecarbaldehyde, n-formylmorfolin czech, unii-d1e63xo4rh, n-formyhnorpholine, n-formyl morpholin
PubChem CID 20417
ChEBI CHEBI:43989
IUPAC Name morpholine-4-carbaldehyde
SMILES C1COCCN1C=O

Specifications

Melting Point 20°C to 23°C
Density 1.15
pH 10
Boiling Point 239°C to 241°C
Flash Point 118°C (244°F)
Refractive Index 1.486
Quantity 100 g
Beilstein 110293
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 115.13
Percent Purity 99%
Chemical Name or Material 4-Formylmorpholine
Show More Show Less
Hazard Category H315-H320
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P501c
EINECSNumber 224-518-3
RTECSNumber QD9694000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only