Learn More
4-Formylcinnamic acid, 98%, predominantly trans
CAS: 23359-08-2 | C10H8O3 | 176.17 g/mol
$290.47 - $290.47
Chemical Identifiers
| CAS | 23359-08-2 |
|---|---|
| Molecular Formula | C10H8O3 |
| Molecular Weight (g/mol) | 176.17 |
| MDL Number | MFCD00006955 |
| InChI Key | LBOUHDMYVURTMA-AATRIKPKSA-N |
| Synonym | 4-formylcinnamic acid, 3-4-formylphenyl acrylic acid, p-formylcinnamic acid, e-3-4-formylphenyl acrylic acid, 2e-3-4-formylphenyl prop-2-enoic acid, 4-2-carboxyvinyl benzaldehyde, p-formylcinnamicacid, 3-4-formylphenyl prop-2-enoic acid, 2e-3-4-formylphenyl acrylic acid, pubchem8244 |
| PubChem CID | 5357280 |
| IUPAC Name | (E)-3-(4-formylphenyl)prop-2-enoic acid |
| SMILES | C1=CC(=CC=C1C=CC(=O)O)C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC176590050
|
Thermo Scientific Chemicals
176590050 |
5 g | Glass bottle |
Each for $290.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 23359-08-2 | |
| 176.17 | |
| LBOUHDMYVURTMA-AATRIKPKSA-N | |
| 5357280 | |
| C1=CC(=CC=C1C=CC(=O)O)C=O |
| C10H8O3 | |
| MFCD00006955 | |
| 4-formylcinnamic acid, 3-4-formylphenyl acrylic acid, p-formylcinnamic acid, e-3-4-formylphenyl acrylic acid, 2e-3-4-formylphenyl prop-2-enoic acid, 4-2-carboxyvinyl benzaldehyde, p-formylcinnamicacid, 3-4-formylphenyl prop-2-enoic acid, 2e-3-4-formylphenyl acrylic acid, pubchem8244 | |
| (E)-3-(4-formylphenyl)prop-2-enoic acid |
Specifications
| 23359-08-2 | |
| 100.0 | |
| Yellow to Beige | |
| 98% | |
| C10H8O3 | |
| MFCD00006955 | |
| 4-formylcinnamic acid, 3-4-formylphenyl acrylic acid, p-formylcinnamic acid, e-3-4-formylphenyl acrylic acid, 2e-3-4-formylphenyl prop-2-enoic acid, 4-2-carboxyvinyl benzaldehyde, p-formylcinnamicacid, 3-4-formylphenyl prop-2-enoic acid, 2e-3-4-formylphenyl acrylic acid, pubchem8244 | |
| C1=CC(=CC=C1C=CC(=O)O)C=O | |
| 176.17 | |
| 176.17 | |
| Crystalline Powder |
| 97.5 | |
| 250°C to 255°C | |
| Authentic | |
| Glass bottle | |
| HCOC6H4CH=CHCO2H | |
| 5 g | |
| LBOUHDMYVURTMA-AATRIKPKSA-N | |
| (E)-3-(4-formylphenyl)prop-2-enoic acid | |
| 5357280 | |
| 98% | |
| 4-Formylcinnamic acid, Predominantly trans |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 245-608-9
RUO – Research Use Only