missing translation for 'onlineSavingsMsg'
Learn More

4'-Formylbenzo-15-crown 5-Ether 98.0+%, TCI America™

$236.73 - $1571.40

Chemical Identifiers

CAS 60835-73-6
Molecular Formula C15H20O6
Molecular Weight (g/mol) 296.319
MDL Number MFCD00192180
InChI Key MBJIKIAWNPEHOR-UHFFFAOYSA-N
Synonym 2,3-(4-Formylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
PubChem CID 324243
IUPAC Name 2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene-17-carbaldehyde
SMILES C1COCCOC2=C(C=C(C=C2)C=O)OCCOCCO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F0448100MG
View Documents
TCI America
F0448100MG
100 mg
Each for $236.73
Only null left
 
F04481G
View Documents
TCI America
F04481G
1 g
Each for $1,571.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

60835-73-6
296.319
MBJIKIAWNPEHOR-UHFFFAOYSA-N
324243
C1COCCOC2=C(C=C(C=C2)C=O)OCCOCCO1
C15H20O6
MFCD00192180
2,3-(4-Formylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene-17-carbaldehyde

Specifications

60835-73-6
C15H20O6
100 mg
MBJIKIAWNPEHOR-UHFFFAOYSA-N
2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene-17-carbaldehyde
324243
≥98.0% (GC)
4′-Formylbenzo-15-crown 5-Ether
85°C
MFCD00192180
2,3-(4-Formylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
C1COCCOC2=C(C=C(C=C2)C=O)OCCOCCO1
296.319
296.32
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'