missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoropyridine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7129794
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06281G 1 g
F06285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06281G Supplier TCI America Supplier No. F06281G
Only null left

Chemical Identifiers

CAS 39160-31-1
Molecular Formula C5H5ClFN
Molecular Weight (g/mol) 133.55
MDL Number MFCD00234055
InChI Key QRUIPALQRYLTAF-UHFFFAOYSA-N
PubChem CID 24820326
IUPAC Name 4-fluoropyridine;hydrochloride
SMILES C1=CN=CC=C1F.Cl

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 1 g
Formula Weight 133.55
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoropyridine Hydrochloride