missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoropyridine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 100°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 133.55 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Fluoropyridine Hydrochloride |
Safety and Handling
| TSCA | No |