missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Fluoropropiophenone 97.0+%, TCI America™
$32.72 - $44.28
Chemical Identifiers
| CAS | 456-03-1 |
|---|---|
| Molecular Formula | C9H9FO |
| Molecular Weight (g/mol) | 152.168 |
| InChI Key | QIJNVLLXIIPXQT-UHFFFAOYSA-N |
| Synonym | 4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone |
| PubChem CID | 68004 |
| IUPAC Name | 1-(4-fluorophenyl)propan-1-one |
| SMILES | CCC(=O)C1=CC=C(C=C1)F |
Chemical Identifiers
| 456-03-1 | |
| 152.168 | |
| 4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone | |
| 1-(4-fluorophenyl)propan-1-one |
| C9H9FO | |
| QIJNVLLXIIPXQT-UHFFFAOYSA-N | |
| 68004 | |
| CCC(=O)C1=CC=C(C=C1)F |
Specifications
| 456-03-1 | |
| 210°C | |
| 5 g | |
| QIJNVLLXIIPXQT-UHFFFAOYSA-N | |
| 1-(4-fluorophenyl)propan-1-one | |
| 68004 | |
| ≥97.0% (GC) | |
| 4′-Fluoropropiophenone |
| Yellow | |
| C9H9FO | |
| 4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone | |
| CCC(=O)C1=CC=C(C=C1)F | |
| 152.168 | |
| 152.17 | |
| Liquid |
Safety and Handling
TSCA : No