missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoropropiophenone 97.0+%, TCI America™

Catalog No. F05225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05225G 5 g
F052225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05225G Supplier TCI America Supplier No. F05225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 456-03-1
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.168
InChI Key QIJNVLLXIIPXQT-UHFFFAOYSA-N
Synonym 4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone
PubChem CID 68004
IUPAC Name 1-(4-fluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)F

Specifications

Color Yellow
Boiling Point 210°C
Quantity 5 g
Formula Weight 152.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Fluoropropiophenone