missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoropropiophenone, 97+%

CAS: 456-03-1 | C9H9FO | 152.168 g/mol

$54.89 - $218.90

Chemical Identifiers

CAS 456-03-1
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.168
MDL Number MFCD00000356
InChI Key QIJNVLLXIIPXQT-UHFFFAOYSA-N
Synonym 4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone
PubChem CID 68004
IUPAC Name 1-(4-fluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1621909
View Documents
Thermo Scientific Chemicals
A1621909
10 g
Each for $54.89
Only null left
 
AAA1621918
View Documents
Thermo Scientific Chemicals
A1621918
50 g
Each for $218.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

456-03-1
152.168
QIJNVLLXIIPXQT-UHFFFAOYSA-N
68004
CCC(=O)C1=CC=C(C=C1)F
C9H9FO
MFCD00000356
4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone
1-(4-fluorophenyl)propan-1-one

Specifications

456-03-1
100°C to 102°C (22 mmHg)
C9H9FO
MFCD00000356
1210310
QIJNVLLXIIPXQT-UHFFFAOYSA-N
1-(4-fluorophenyl)propan-1-one
68004
≥97%
1.096
76°C (168°F)
1.5059
10 g
4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone
CCC(=O)C1=CC=C(C=C1)F
152.168
152.17
4'-Fluoropropiophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 207-255-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only