missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoronaphthalene-1-carbonitrile, 97%

Catalog No. AAB2533503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2533503 1 g
AAB2533506 5 g
AAB2533514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2533503 Supplier Thermo Scientific Chemicals Supplier No. B2533503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13916-99-9 | C11H6FN | 171.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13916-99-9
Molecular Formula C11H6FN
Molecular Weight (g/mol) 171.174
MDL Number MFCD00236656
InChI Key XQSGYYNFKIQWAC-UHFFFAOYSA-N
PubChem CID 2736734
IUPAC Name 4-fluoronaphthalene-1-carbonitrile
SMILES C1=CC=C2C(=C1)C(=CC=C2F)C#N

Specifications

Melting Point 89°C to 91°C
Quantity 1 g
UN Number UN3439
Synonym 1-cyano-4-fluoronaphthalene, 4-fluoro-1-naphthonitrile, 4-fluoro-1-naphthalenecarbonitrile, 1-naphthalenecarbonitrile,4-fluoro, 1-naphthalenecarbonitrile, 4-fluoro, 4-fluoronaphthalenecarbonitrile, 4-fluoro-1-phthonitrile, acmc-1buk6, 1-cyano-4-fluoronapthalene, 4-cyano-1-fluoronaphthalene
Formula Weight 171.18
Percent Purity 97%
Chemical Name or Material 4-Fluoronaphthalene-1-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only