missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoroisobutyrophenone, 97%

Catalog No. AAH2603603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2603603 1 g
AAH2603606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2603603 Supplier Thermo Scientific Chemicals Supplier No. H2603603
Only null left

CAS: 26393-91-9 | C10H11FO | 166.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26393-91-9
Molecular Formula C10H11FO
Molecular Weight (g/mol) 166.20
MDL Number MFCD04038305
InChI Key MHUVRVXSYXJUPK-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl-2-methylpropan-1-one, 4'-fluoroisobutyrophenone, 1-4-fluorophenyl-2-methyl-1-propanone, 1-4-fluoro-phenyl-2-methyl-propan-1-one, 1-propanone, 1-4-fluorophenyl-2-methyl, p-fluoroisobutyrophenone, 1-4-fluorophenyl-2-methyl-propan-1-one, acmc-1chy0, isopropyl p-fluorophenyl ketone, p-fluorophenyl isopropyl ketone
PubChem CID 2782829
IUPAC Name 1-(4-fluorophenyl)-2-methylpropan-1-one
SMILES CC(C)C(=O)C1=CC=C(F)C=C1

Specifications

Refractive Index 1.498
Quantity 1 g
Formula Weight 166.19
Percent Purity 97%
Chemical Name or Material 4'-Fluoroisobutyrophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only