missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroindole 98.0+%, TCI America™

Catalog No. F05561G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05561G Supplier TCI America Supplier No. F05561G

Chemical Identifiers

CAS 387-43-9
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD00055992
InChI Key ZWKIJOPJWWZLDI-UHFFFAOYSA-N
Synonym 4-fluoroindole, 1h-indole, 4-fluoro, 4-fluoro-indole, pubchem7297, acmc-1cnb8, ksc224m5j, bidd:gt0103
PubChem CID 2774502
IUPAC Name 4-fluoro-1H-indole
SMILES C1=CC2=C(C=CN2)C(=C1)F

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 90°C
Quantity 1 g
Formula Weight 135.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoroindole