missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroindole, 96%

Catalog No. AC397650010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397650010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC397650010 Supplier Thermo Scientific Chemicals Supplier No. 397650010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 387-43-9 | C8H6FN | 135.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 387-43-9
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00055992
InChI Key ZWKIJOPJWWZLDI-UHFFFAOYSA-N
Synonym 4-fluoroindole, 1h-indole, 4-fluoro, 4-fluoro-indole, pubchem7297, acmc-1cnb8, ksc224m5j, bidd:gt0103
PubChem CID 2774502
IUPAC Name 4-fluoro-1H-indole
SMILES C1=CC2=C(C=CN2)C(=C1)F

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 32°C
Color Tan to Yellow
Boiling Point 90°C (0.4 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 135.14
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoroindole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only