Learn More
4-Fluorocinnamic acid, 98+%
CAS: 459-32-5 | C9H7FO2 | 166.151 g/mol
Supplier: Thermo Scientific Chemicals A1670706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Fluorocinnamic acid | |
| 208°C to 210°C | |
| C9H7FO2 | |
| 5 g | |
| 4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e | |
| C1=CC(=CC=C1C=CC(=O)O)F | |
| 166.151 | |
| 166.15 |
| 459-32-5 | |
| Odorless | |
| MFCD00004395 | |
| 2613441 | |
| ISMMYAZSUSYVQG-ZZXKWVIFSA-N | |
| (E)-3-(4-fluorophenyl)prop-2-enoic acid | |
| 1530234 | |
| ≥98% |
Chemical Identifiers
| 459-32-5 | |
| 166.151 | |
| ISMMYAZSUSYVQG-ZZXKWVIFSA-N | |
| 1530234 | |
| C1=CC(=CC=C1C=CC(=O)O)F |
| C9H7FO2 | |
| MFCD00004395 | |
| 4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e | |
| (E)-3-(4-fluorophenyl)prop-2-enoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-288-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only