missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluorobiphenyl-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog No. p-4813118
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450320050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC450320050 Supplier Thermo Scientific Chemicals Supplier No. 450320050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 116748-66-4
Molecular Formula C12H8ClFO2S
Molecular Weight (g/mol) 270.71
InChI Key CIDMHDJTWVMBIF-UHFFFAOYSA-N
Synonym 4'-fluorobiphenyl-4-sulfonyl chloride, 4-4-fluorophenyl benzenesulfonyl chloride, 4'-fluoro 1,1'-biphenyl-4-sulfonyl chloride, 4'-fluoro-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-fluoro, 4'-fluoro-1,1'-biphenyl-4-sulfonyl chloride, acmc-20apun, 4'-fluorobiphenyl-4-sulfonylchloride, 4'-fluoro-4-biphenylsulfonyl chloride
PubChem CID 2794749
IUPAC Name 4-(4-fluorophenyl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)S(=O)(=O)Cl)F

Specifications

Melting Point 82.0°C to 84.0°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 270.71
Percent Purity 97%
Chemical Name or Material 4'-Fluorobiphenyl-4-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

RUO – Research Use Only