missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl mercaptan, 96%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 443900010

Catalog No. AC443900010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

 

Specifications

Specifications

4-Fluorobenzyl mercaptan
95.0
1.1570g/mL
76°C
95% min. (GC)
C7H7FS
1.157
RKTRHMNWVZRZJQ-UHFFFAOYSA-N
(4-fluorophenyl)methanethiol
85171
96%
15894-04-9
100.0
72°C to 74°C (15.0mmHg)
Authentic
Glass Bottle
1g
4-fluorobenzyl mercaptan, 4-fluorophenyl methanethiol, 4-fluorobenzylmercaptan, 4-fluoro benzyl mercaptan, 4-fluorobenzenemethanethiol, p-fluorotoluene-alpha-thiol, benzenemethanethiol, 4-fluoro, 4-fluoro-phenyl-methanethiol, 4-fluorobenzylthiol, 4-fluorobenzyl thiol
C1=CC(=CC=C1CS)F
142.2
142.2

Chemical Identifiers

15894-04-9
142.2
4-fluorobenzyl mercaptan, 4-fluorophenyl methanethiol, 4-fluorobenzylmercaptan, 4-fluoro benzyl mercaptan, 4-fluorobenzenemethanethiol, p-fluorotoluene-alpha-thiol, benzenemethanethiol, 4-fluoro, 4-fluoro-phenyl-methanethiol, 4-fluorobenzylthiol, 4-fluorobenzyl thiol
(4-fluorophenyl)methanethiol
C7H7FS
RKTRHMNWVZRZJQ-UHFFFAOYSA-N
85171
C1=CC(=CC=C1CS)F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 240-031-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only