missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoylacetonitrile 98.0+%, TCI America™

Catalog No. F09875G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09875G 5 g
F09871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09875G Supplier TCI America Supplier No. F09875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4640-67-9
Molecular Formula C9H6FNO
Molecular Weight (g/mol) 163.15
MDL Number MFCD00662062
InChI Key LOJBBLDAJBJVBZ-UHFFFAOYSA-N
Synonym 4-fluorobenzoylacetonitrile, 3-4-fluorophenyl-3-oxopropanenitrile, p-fluorobenzoylacetonitrile, unii-k1ltb8k2dh, 3-4-fluorophenyl-3-oxopropionitrile, k1ltb8k2dh, 3-4-fluoro-phenyl-3-oxo-propionitrile, benzenepropanenitrile, 4-fluoro-beta-oxo, alpha-cyano-4-fluoroacetophenone, acetonitrile, p-fluorobenzoyl
PubChem CID 2783172
IUPAC Name 3-(4-fluorophenyl)-3-oxopropanenitrile
SMILES FC1=CC=C(C=C1)C(=O)CC#N

Specifications

Melting Point 85°C
Color Yellow
Quantity 5 g
Formula Weight 163.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzoylacetonitrile