missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzenesulfonyl Chloride 97.0+%, TCI America™

Catalog No. F02025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02025G 5 g
F020225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02025G Supplier TCI America Supplier No. F02025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 349-88-2
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.60
MDL Number MFCD00007438
InChI Key BFXHJFKKRGVUMU-UHFFFAOYSA-N
Synonym 4-fluorobenzene-1-sulfonyl chloride, p-fluorobenzenesulfonyl chloride, 4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro, 4-fluorophenylsulfonyl chloride, 4-fluorobenzenesulfonic acid chloride, 4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, p-fluoro, 4-fluorobenzensulfonyl chloride, 4-fluoro-benzenesulfonyl chloride
PubChem CID 9588
IUPAC Name 4-fluorobenzene-1-sulfonyl chloride
SMILES FC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 35°C
Color White
Boiling Point 112°C
Quantity 5 g
UN Number 3261
Formula Weight 194.60
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Fluorobenzenesulfonyl Chloride