missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzenesulfonyl chloride, 98%

Catalog No. p-3778595
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119350250 25 g Glass bottle
AC119351000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC119350250 Supplier Thermo Scientific Chemicals Supplier No. 119350250

CAS: 349-88-2 | C6H4ClFO2S | 194.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 349-88-2
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.60
MDL Number MFCD00007438
InChI Key BFXHJFKKRGVUMU-UHFFFAOYSA-N
Synonym 4-fluorobenzene-1-sulfonyl chloride, p-fluorobenzenesulfonyl chloride, 4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro, 4-fluorophenylsulfonyl chloride, 4-fluorobenzenesulfonic acid chloride, 4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, p-fluoro, 4-fluorobenzensulfonyl chloride, 4-fluoro-benzenesulfonyl chloride
PubChem CID 9588
IUPAC Name 4-fluorobenzenesulfonyl chloride
SMILES FC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29°C to 34°C
Color Brown to White
Boiling Point 95°C to 96°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula FC6H4SO2Cl
Quantity 25 g
Beilstein 11, 53
Formula Weight 194.61
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Fluorobenzenesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 206-493-

RUO – Research Use Only