Learn More
4-Fluorobenzenesulfonyl chloride, 98%
CAS: 349-88-2 | C6H4ClFO2S | 194.60 g/mol
Supplier: Thermo Scientific Chemicals L0729914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Fluorobenzenesulfonyl chloride | |
| 35°C to 38°C | |
| >110°C (230°F) | |
| MFCD00007438 | |
| UN3261 | |
| Moisture sensitive | |
| BFXHJFKKRGVUMU-UHFFFAOYSA-N | |
| 4-fluorobenzenesulfonyl chloride | |
| 9588 | |
| 98% |
| 349-88-2 | |
| 86°C to 87°C (1 mmHg) | |
| C6H4ClFO2S | |
| 25 g | |
| 2091633 | |
| 4-fluorobenzene-1-sulfonyl chloride, p-fluorobenzenesulfonyl chloride, 4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro, 4-fluorophenylsulfonyl chloride, 4-fluorobenzenesulfonic acid chloride, 4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, p-fluoro, 4-fluorobenzensulfonyl chloride, 4-fluoro-benzenesulfonyl chloride | |
| FC1=CC=C(C=C1)S(Cl)(=O)=O | |
| 194.60 | |
| 194.61 |
Chemical Identifiers
| 349-88-2 | |
| 194.60 | |
| BFXHJFKKRGVUMU-UHFFFAOYSA-N | |
| 9588 | |
| FC1=CC=C(C=C1)S(Cl)(=O)=O |
| C6H4ClFO2S | |
| MFCD00007438 | |
| 4-fluorobenzene-1-sulfonyl chloride, p-fluorobenzenesulfonyl chloride, 4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro, 4-fluorophenylsulfonyl chloride, 4-fluorobenzenesulfonic acid chloride, 4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, p-fluoro, 4-fluorobenzensulfonyl chloride, 4-fluoro-benzenesulfonyl chloride | |
| 4-fluorobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 206-493-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only