missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzaldehyde, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 156150010

Catalog No. AC156150010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Fluorobenzaldehyde
459-57-4
100.0
181.0°C (758.0 mmHg)
Authentic
Glass bottle
1.5195 to 1.5215
MFCD00003378
07, I, 132
p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde
UOQXIWFBQSVDPP-UHFFFAOYSA-N
4-fluorobenzaldehyde
68023
98+%
98+%
98.0
1.1500g/mL
56°C
98% min. (GC)
C7H5FO
FC6H4CHO
1L
1.15
Solubility in water: immiscible. Other solubilities: soluble in ether,acetone,benzene and alcohol
C1=CC(=CC=C1C=O)F
124.11
124.11
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-293-6

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only