missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoroacetophenone 98.0+%, TCI America™

Catalog No. F0261500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0261500G 500 g
F026125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0261500G Supplier TCI America Supplier No. F0261500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 403-42-9
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.14
MDL Number MFCD00000354
InChI Key ZDPAWHACYDRYIW-UHFFFAOYSA-N
Synonym 4'-fluoroacetophenone, 4-fluoroacetophenone, 1-4-fluorophenyl ethanone, p-fluoroacetophenone, 1-4-fluorophenyl ethan-1-one, ethanone, 1-4-fluorophenyl, acetophenone, 4'-fluoro, 4 fluoroacetophenone, unii-tvq090602v, 1-acetyl-4-fluorobenzene
PubChem CID 9828
IUPAC Name 1-(4-fluorophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(F)C=C1

Specifications

Color Yellow
Boiling Point 77°C
Quantity 500 g
Formula Weight 138.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Fluoroacetophenone