missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoroacetophenone, 99%

Catalog No. AAA1395014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1395014 25 g
AAA1395022 100 g
AAA1395036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1395014 Supplier Thermo Scientific Chemicals Supplier No. A1395014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 403-42-9 | C8H7FO | 138.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 403-42-9
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.14
MDL Number MFCD00000354
InChI Key ZDPAWHACYDRYIW-UHFFFAOYSA-N
Synonym 4'-fluoroacetophenone, 4-fluoroacetophenone, 1-4-fluorophenyl ethanone, p-fluoroacetophenone, 1-4-fluorophenyl ethan-1-one, ethanone, 1-4-fluorophenyl, acetophenone, 4'-fluoro, 4 fluoroacetophenone, unii-tvq090602v, 1-acetyl-4-fluorobenzene
PubChem CID 9828
IUPAC Name 1-(4-fluorophenyl)ethanone
SMILES CC(=O)C1=CC=C(F)C=C1

Specifications

Melting Point 4°C
Density 1.141
Boiling Point 194°C to 196°C
Flash Point 71°C (159°F)
Refractive Index 1.511
Quantity 25 g
Beilstein 386013
Formula Weight 138.14
Percent Purity 99%
Chemical Name or Material 4'-Fluoroacetophenone
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-960-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only