missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N-salicylideneaniline 98.0+%, TCI America™

Catalog No. F08935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08935G 5 g
F08931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08935G Supplier TCI America Supplier No. F08935G
Only null left

Chemical Identifiers

CAS 3382-62-5
Molecular Formula C13H10FNO
Molecular Weight (g/mol) 215.227
MDL Number MFCD00020090
InChI Key IOQIIBHVEXJHMV-UHFFFAOYSA-N
Synonym 4-Fluoro-N-salicylalaniline
PubChem CID 6739742
IUPAC Name 6-[(4-fluoroanilino)methylidene]cyclohexa-2,4-dien-1-one
SMILES C1=CC(=CNC2=CC=C(C=C2)F)C(=O)C=C1

Specifications

Melting Point 84°C
Color Yellow
Quantity 5 g
Formula Weight 215.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-N-salicylideneaniline