missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-D-phenylalanine 98.0+%, TCI America™
$119.30 - $352.30
Chemical Identifiers
| CAS | 18125-46-7 |
|---|---|
| Molecular Formula | C9H10FNO2 |
| Molecular Weight (g/mol) | 183.182 |
| MDL Number | MFCD00063063 |
| InChI Key | XWHHYOYVRVGJJY-MRVPVSSYSA-N |
| Synonym | 4-fluoro-d-phenylalanine, d-4-fluorophenylalanine, h-d-phe 4-f-oh, r-2-amino-3-4-fluorophenyl propanoic acid, p-fluoro-d-phenylalanine, d-phenylalanine, 4-fluoro, 2r-2-amino-3-4-fluorophenyl propanoic acid, 4-fluorophenylalanine, d, unii-963g7o1pve, r-4-fluorophenylalanine |
| PubChem CID | 716314 |
| IUPAC Name | (2R)-2-amino-3-(4-fluorophenyl)propanoic acid |
| SMILES | C1=CC(=CC=C1CC(C(=O)O)N)F |
Chemical Identifiers
| 18125-46-7 | |
| 183.182 | |
| XWHHYOYVRVGJJY-MRVPVSSYSA-N | |
| 716314 | |
| C1=CC(=CC=C1CC(C(=O)O)N)F |
| C9H10FNO2 | |
| MFCD00063063 | |
| 4-fluoro-d-phenylalanine, d-4-fluorophenylalanine, h-d-phe 4-f-oh, r-2-amino-3-4-fluorophenyl propanoic acid, p-fluoro-d-phenylalanine, d-phenylalanine, 4-fluoro, 2r-2-amino-3-4-fluorophenyl propanoic acid, 4-fluorophenylalanine, d, unii-963g7o1pve, r-4-fluorophenylalanine | |
| (2R)-2-amino-3-(4-fluorophenyl)propanoic acid |
Specifications
| 18125-46-7 | |
| White | |
| MFCD00063063 | |
| 4-fluoro-d-phenylalanine, d-4-fluorophenylalanine, h-d-phe 4-f-oh, r-2-amino-3-4-fluorophenyl propanoic acid, p-fluoro-d-phenylalanine, d-phenylalanine, 4-fluoro, 2r-2-amino-3-4-fluorophenyl propanoic acid, 4-fluorophenylalanine, d, unii-963g7o1pve, r-4-fluorophenylalanine | |
| C1=CC(=CC=C1CC(C(=O)O)N)F | |
| 183.182 | |
| 183.18 | |
| Crystalline Powder |
| 247°C | |
| C9H10FNO2 | |
| 1 g | |
| XWHHYOYVRVGJJY-MRVPVSSYSA-N | |
| (2R)-2-amino-3-(4-fluorophenyl)propanoic acid | |
| 716314 | |
| ≥98.0% (HPLC,T) | |
| 4-Fluoro-D-phenylalanine |
Safety and Handling
TSCA : No