missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-4'-methylchalcone, TCI America™

Catalog No. F04605G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04605G Supplier TCI America Supplier No. F04605G
Only null left

Chemical Identifiers

CAS 13565-38-3
Molecular Formula C16H13FO
Molecular Weight (g/mol) 240.277
MDL Number MFCD00017981
InChI Key LUUPODNGYDYQLY-IZZDOVSWSA-N
Synonym 4-fluoro-4'-methylchalcone, 2e-3-4-fluorophenyl-1-4-methylphenyl prop-2-en-1-one, e-3-4-fluorophenyl-1-p-tolyl prop-2-en-1-one, e-3-4-fluorophenyl-1-4-methylphenyl prop-2-en-1-one, e-4-fluoro-4'-methylchalcone, 2z-3-4-fluorophenyl-1-4-methylphenyl-2-propen-1-one, e-3-4-fluorophenyl-1-4-methylphenyl-2-propen-1-one
PubChem CID 5702626
IUPAC Name (E)-3-(4-fluorophenyl)-1-(4-methylphenyl)prop-2-en-1-one
SMILES CC1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 240.28
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-4′-methylchalcone