missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-3'-(trifluoromethyl)acetophenone 97.0+%, TCI America™

Catalog No. F08085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08085G 5 g
F08081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08085G Supplier TCI America Supplier No. F08085G
Only null left

Chemical Identifiers

CAS 208173-24-4
Molecular Formula C9H6F4O
Molecular Weight (g/mol) 206.14
MDL Number MFCD00061252
InChI Key SYFHRXQPXHETEF-UHFFFAOYSA-N
Synonym 4'-fluoro-3'-trifluoromethyl acetophenone, 1-4-fluoro-3-trifluoromethyl phenyl ethanone, 4-fluoro-3-trifluoromethyl acetophenone, 4'-fluoro-3'-trifluoromethylacetophenone, 1-4-fluoro-3-trifluoromethyl phenyl ethan-1-one, 5-acetyl-2-fluorobenzotrifluoride, ethanone, 1-4-fluoro-3-trifluoromethyl phenyl, 1-acetyl-4-fluoro-3-trifluoromethyl benzene, pubchem4321
PubChem CID 605678
IUPAC Name 1-[4-fluoro-3-(trifluoromethyl)phenyl]ethan-1-one
SMILES CC(=O)C1=CC=C(F)C(=C1)C(F)(F)F

Specifications

Color Colorless
Quantity 5 g
Formula Weight 206.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Fluoro-3′-(trifluoromethyl)acetophenone