missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid, 95%, Thermo Scientific™

Chemical Identifiers

CAS 1009303-56-3
Molecular Formula C7H6BFN4O2
Molecular Weight (g/mol) 207.96
MDL Number MFCD13176533
InChI Key FELFJHZMQYCKPT-UHFFFAOYSA-N
Synonym 4-fluoro-3-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenyl boronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenylboronic acid
PubChem CID 11287236
IUPAC Name [4-fluoro-3-(2H-tetrazol-5-yl)phenyl]boronic acid
SMILES OB(O)C1=CC(C2=NNN=N2)=C(F)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445460010
SDS View Documents
Thermo Scientific Chemicals
445460010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1009303-56-3
207.96
FELFJHZMQYCKPT-UHFFFAOYSA-N
11287236
OB(O)C1=CC(C2=NNN=N2)=C(F)C=C1
C7H6BFN4O2
MFCD13176533
4-fluoro-3-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenyl boronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenylboronic acid
[4-fluoro-3-(2H-tetrazol-5-yl)phenyl]boronic acid

Specifications

1009303-56-3
Glass Bottle
MFCD13176533
4-fluoro-3-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenyl boronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenylboronic acid
OB(O)C1=CC(C2=NNN=N2)=C(F)C=C1
207.96
207.96
4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid
94% min. (HPLC)
C7H6BFN4O2
1g
FELFJHZMQYCKPT-UHFFFAOYSA-N
[4-fluoro-3-(2H-tetrazol-5-yl)phenyl]boronic acid
11287236
95%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only