missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 305060250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC305060250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

453-71-4
185.11
BOJWTAQWPVBIPG-UHFFFAOYSA-N
67987
OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
C7H4FNO4
MFCD00007058
4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid
4-fluoro-3-nitrobenzoic acid

Specifications

4-Fluoro-3-nitrobenzoic acid
453-71-4
100.0
97.5% min. (GC)
C7H4FNO4
25g
4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid
BOJWTAQWPVBIPG-UHFFFAOYSA-N
4-fluoro-3-nitrobenzoic acid
67987
98%
98%
97.5
Authentic
Glass bottle
FC6H3(NO2)CO2H
MFCD00007058
Solubility in water: insoluble in water
OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
185.11
185.11
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-221-3

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...