missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-3'-nitroacetophenone 98.0+%, TCI America™

Catalog No. F07775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07775G 5 g
F077725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07775G Supplier TCI America Supplier No. F07775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 400-93-1
Molecular Formula C8H6FNO3
Molecular Weight (g/mol) 183.138
MDL Number MFCD00115369
InChI Key PTCNZDJJIOLIKQ-UHFFFAOYSA-N
Synonym 4'-fluoro-3'-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethanone, 4-fluoro-3-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethan-1-one, 1-4-fluoro-3-nitro-phenyl ethanone, 4-fluoro-3'-nitroacetophenone, ethanone, 1-4-fluoro-3-nitrophenyl, 4-fluoro-3-nitroacetophneone, 3'-nitro-4'-fluoroacetophenone
PubChem CID 2734612
IUPAC Name 1-(4-fluoro-3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-]

Specifications

Melting Point 50°C
Color Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 183.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Fluoro-3′-nitroacetophenone