missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7193790
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09191G Supplier TCI America Supplier No. F09191G

Chemical Identifiers

CAS 854778-31-7
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
MDL Number MFCD08056358
InChI Key LUJMSRVFSBMEOY-UHFFFAOYSA-N
PubChem CID 44558185
IUPAC Name (4-fluoro-3-methoxyphenyl)boronic acid
SMILES COC1=C(F)C=CC(=C1)B(O)O

Specifications

Melting Point 191°C
Color White
Quantity 1 g
Synonym 4-fluoro-3-methoxyphenyl boronic acid, 4-fluoro-3-methoxybenzeneboronic acid, boronic acid, 4-fluoro-3-methoxyphenyl, ksc495c1j, 3-methoxy4-fluorophenylboronic acid, 4-fluoro-3-methoxyphenylboronicacid, 3-methoxy-4-fluorophenylboronic acid, 4-fluoro-5-methoxyphenylboronic acid, boronic acid,b-4-fluoro-3-methoxyphenyl
Formula Weight 169.95
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-methoxyphenylboronic Acid (contains varying amounts of Anhydride)