missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-2-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
$538.42 - $538.42
Chemical Identifiers
| CAS | 1288978-82-4 |
|---|---|
| Molecular Formula | C12H15BFNO4 |
| Molecular Weight (g/mol) | 267.06 |
| MDL Number | MFCD16996295 |
| InChI Key | UEMDKWBOUAQJJU-UHFFFAOYSA-N |
| PubChem CID | 71744276 |
| IUPAC Name | 2-(4-fluoro-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| SMILES | CC1(C)OB(OC1(C)C)C1=C(C=C(F)C=C1)[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6205903
|
Thermo Scientific Chemicals
H6205903 |
1 g |
Each for $538.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1288978-82-4 | |
| 267.06 | |
| UEMDKWBOUAQJJU-UHFFFAOYSA-N | |
| 2-(4-fluoro-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| C12H15BFNO4 | |
| MFCD16996295 | |
| 71744276 | |
| CC1(C)OB(OC1(C)C)C1=C(C=C(F)C=C1)[N+]([O-])=O |
Specifications
| 1288978-82-4 | |
| MFCD16996295 | |
| 2-4-fluoro-2-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-nitrobenzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-fluoro-2-nitrophenyl-4,4,5,5-tetramethyl, 1,3,2-dioxaborolane,2-4-fluoro-2-nitrophenyl-4,4,5,5-tetramethyl, 4-fluoro-2-nitrophenylboronic acid pinacol ester, 4-fluoro-2-nitrophenyl boronic acid pinacol ester | |
| CC1(C)OB(OC1(C)C)C1=C(C=C(F)C=C1)[N+]([O-])=O | |
| 267.06 | |
| 267.06 | |
| Solid |
| C12H15BFNO4 | |
| 1 g | |
| UEMDKWBOUAQJJU-UHFFFAOYSA-N | |
| 2-(4-fluoro-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 71744276 | |
| 96% | |
| 4-Fluoro-2-nitrobenzeneboronic acid pinacol ester |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only