missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitroaniline 98.0+%, TCI America™

Catalog No. F03805G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03805G 5 g
F038025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03805G Supplier TCI America Supplier No. F03805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 364-78-3
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00007830
InChI Key PUGDHSSOXPHLPT-UHFFFAOYSA-N
Synonym 2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421
PubChem CID 67769
IUPAC Name 4-fluoro-2-nitroaniline
SMILES NC1=CC=C(F)C=C1[N+]([O-])=O

Specifications

Melting Point 94°C
Quantity 5 g
Formula Weight 156.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-nitroaniline