Learn More
4-Fluoro-2-nitroaniline, 99%
CAS: 364-78-3 | C6H5FN2O2 | 156.12 g/mol
$56.86 - $56.86
Chemical Identifiers
| CAS | 364-78-3 |
|---|---|
| Molecular Formula | C6H5FN2O2 |
| Molecular Weight (g/mol) | 156.12 |
| MDL Number | MFCD00007830 |
| InChI Key | PUGDHSSOXPHLPT-UHFFFAOYSA-N |
| Synonym | 2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421 |
| PubChem CID | 67769 |
| IUPAC Name | 4-fluoro-2-nitroaniline |
| SMILES | NC1=CC=C(F)C=C1[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC233070100
|
Thermo Scientific Chemicals
233070100 |
10 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 364-78-3 | |
| 156.12 | |
| PUGDHSSOXPHLPT-UHFFFAOYSA-N | |
| 67769 | |
| NC1=CC=C(F)C=C1[N+]([O-])=O |
| C6H5FN2O2 | |
| MFCD00007830 | |
| 2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421 | |
| 4-fluoro-2-nitroaniline |
Specifications
| 364-78-3 | |
| 100.0 | |
| Orange to Brown | |
| Authentic | |
| Glass bottle | |
| FC6H3(NO2)NH2 | |
| 10 g | |
| Solubility in water: insoluble. Other solubilities: soluble in polar organic solvents,insoluble in non-polar organic solvents,approx. 12% soluble in alcohol (15°C) | |
| NC1=CC=C(F)C=C1[N+]([O-])=O | |
| 156.12 | |
| 156.12 | |
| Crystalline Powder |
| 98.5 | |
| 92°C to 95°C | |
| 89°C | |
| 99% | |
| C6H5FN2O2 | |
| MFCD00007830 | |
| 2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421 | |
| PUGDHSSOXPHLPT-UHFFFAOYSA-N | |
| 4-fluoro-2-nitroaniline | |
| 67769 | |
| 99% | |
| 4-Fluoro-2-nitroaniline, 99% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Flammable solid.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 206-666-0
RUO – Research Use Only