missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitroaniline, 99%

Catalog No. AC233070100
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC233070100 10 g Glass bottle
AC233071000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC233070100 Supplier Thermo Scientific Chemicals Supplier No. 233070100

CAS: 364-78-3 | C6H5FN2O2 | 156.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 364-78-3
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00007830
InChI Key PUGDHSSOXPHLPT-UHFFFAOYSA-N
Synonym 2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421
PubChem CID 67769
IUPAC Name 4-fluoro-2-nitroaniline
SMILES NC1=CC=C(F)C=C1[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 92°C to 95°C
Color Orange to Brown
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula FC6H3(NO2)NH2
Quantity 10 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in polar organic solvents,insoluble in non-polar organic solvents,approx. 12% soluble in alcohol (15°C)
Formula Weight 156.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-nitroaniline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Flammable solid.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 206-666-0

RUO – Research Use Only