Learn More
4-Fluoro-2-methylbenzenesulfonyl chloride, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 7079-48-3 |
|---|---|
| Molecular Formula | C7H6ClFO2S |
| Molecular Weight (g/mol) | 208.64 |
| InChI Key | XLPGWKNCWMFHOD-UHFFFAOYSA-N |
| Synonym | 4-fluoro-2-methylbenzene-1-sulfonyl chloride, 4-fluoro-2-methylbenzenesulfonylchloride, 4-fluoro-2-methylbenzenesulphonyl chloride, 4-fluoro-2-methyl-benzenesulfonyl chloride, chloro 4-fluoro-2-methylphenyl sulfone, pubchem11719, ablock ab-12-8862, 4-fluoro-2-methylphenylsulfonyl chloride, 4-fluoro-2-methylbenzene-1-sulfonylchloride, benzenesulfonylchloride, 4-fluoro-2-methyl |
| PubChem CID | 2778602 |
| IUPAC Name | 4-fluoro-2-methylbenzenesulfonyl chloride |
| SMILES | CC1=C(C=CC(=C1)F)S(=O)(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC442370050
|
Thermo Scientific Chemicals
442370050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 7079-48-3 | |
| 208.64 | |
| 4-fluoro-2-methylbenzene-1-sulfonyl chloride, 4-fluoro-2-methylbenzenesulfonylchloride, 4-fluoro-2-methylbenzenesulphonyl chloride, 4-fluoro-2-methyl-benzenesulfonyl chloride, chloro 4-fluoro-2-methylphenyl sulfone, pubchem11719, ablock ab-12-8862, 4-fluoro-2-methylphenylsulfonyl chloride, 4-fluoro-2-methylbenzene-1-sulfonylchloride, benzenesulfonylchloride, 4-fluoro-2-methyl | |
| 4-fluoro-2-methylbenzenesulfonyl chloride |
| C7H6ClFO2S | |
| XLPGWKNCWMFHOD-UHFFFAOYSA-N | |
| 2778602 | |
| CC1=C(C=CC(=C1)F)S(=O)(=O)Cl |
Specifications
| 7079-48-3 | |
| 100.0 | |
| 110°C | |
| 96% min. (GC) | |
| 5g | |
| XLPGWKNCWMFHOD-UHFFFAOYSA-N | |
| 4-fluoro-2-methylbenzenesulfonyl chloride | |
| 2778602 | |
| 97% |
| 96.0 | |
| 150°C to 152°C (92.0 Torr) | |
| Authentic | |
| C7H6ClFO2S | |
| 4-fluoro-2-methylbenzene-1-sulfonyl chloride, 4-fluoro-2-methylbenzenesulfonylchloride, 4-fluoro-2-methylbenzenesulphonyl chloride, 4-fluoro-2-methyl-benzenesulfonyl chloride, chloro 4-fluoro-2-methylphenyl sulfone, pubchem11719, ablock ab-12-8862, 4-fluoro-2-methylphenylsulfonyl chloride, 4-fluoro-2-methylbenzene-1-sulfonylchloride, benzenesulfonylchloride, 4-fluoro-2-methyl | |
| CC1=C(C=CC(=C1)F)S(=O)(=O)Cl | |
| 208.64 | |
| 208.64 | |
| 4-Fluoro-2-methylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO â Research Use Only