missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog No. AC442370050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442370050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC442370050 Supplier Thermo Scientific Chemicals Supplier No. 442370050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7079-48-3 | Molecular Formula: C7H6ClFO2S | Molecular Weight: 208.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7079-48-3
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.64
InChI Key XLPGWKNCWMFHOD-UHFFFAOYSA-N
Synonym 4-fluoro-2-methylbenzene-1-sulfonyl chloride, 4-fluoro-2-methylbenzenesulfonylchloride, 4-fluoro-2-methylbenzenesulphonyl chloride, 4-fluoro-2-methyl-benzenesulfonyl chloride, chloro 4-fluoro-2-methylphenyl sulfone, pubchem11719, ablock ab-12-8862, 4-fluoro-2-methylphenylsulfonyl chloride, 4-fluoro-2-methylbenzene-1-sulfonylchloride, benzenesulfonylchloride, 4-fluoro-2-methyl
PubChem CID 2778602
IUPAC Name 4-fluoro-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=CC(=C1)F)S(=O)(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 150°C to 152°C (92.0 Torr)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 208.64
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-methylbenzenesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only