missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzenesulfonyl chloride, 97%

Catalog No. AAL1951706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1951706 5 g
AAL1951703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1951706 Supplier Thermo Scientific Chemicals Supplier No. L1951706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7079-48-3 | C7H6ClFO2S | 208.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7079-48-3
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.631
MDL Number MFCD03094390
InChI Key XLPGWKNCWMFHOD-UHFFFAOYSA-N
Synonym 4-fluoro-2-methylbenzene-1-sulfonyl chloride, 4-fluoro-2-methylbenzenesulfonylchloride, 4-fluoro-2-methylbenzenesulphonyl chloride, 4-fluoro-2-methyl-benzenesulfonyl chloride, chloro 4-fluoro-2-methylphenyl sulfone, pubchem11719, ablock ab-12-8862, 4-fluoro-2-methylphenylsulfonyl chloride, 4-fluoro-2-methylbenzene-1-sulfonylchloride, benzenesulfonylchloride, 4-fluoro-2-methyl
PubChem CID 2778602
IUPAC Name 4-fluoro-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=CC(=C1)F)S(=O)(=O)Cl

Specifications

Density 1.433
Boiling Point 245°C to 246°C
Flash Point >110°C (230°F)
Refractive Index 1.537
Quantity 5 g
UN Number UN3265
Beilstein 6720115
Sensitivity Moisture sensitive
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylbenzenesulfonyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only