missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$156.59 - $156.59
Chemical Identifiers
| CAS | 179899-07-1 |
|---|---|
| Molecular Formula | C7H8BFO3 |
| Molecular Weight (g/mol) | 169.95 |
| MDL Number | MFCD03788423 |
| InChI Key | ADJBXDCXYMCCAD-UHFFFAOYSA-N |
| PubChem CID | 2782672 |
| IUPAC Name | (4-fluoro-2-methoxyphenyl)boronic acid |
| SMILES | COC1=CC(F)=CC=C1B(O)O |
Chemical Identifiers
| 179899-07-1 | |
| 169.95 | |
| ADJBXDCXYMCCAD-UHFFFAOYSA-N | |
| (4-fluoro-2-methoxyphenyl)boronic acid |
| C7H8BFO3 | |
| MFCD03788423 | |
| 2782672 | |
| COC1=CC(F)=CC=C1B(O)O |
Specifications
| 179899-07-1 | |
| White | |
| MFCD03788423 | |
| 4-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-4-fluorophenylboronic acid, 4-fluoro-2-methoxybenzeneboronic acid, 4-fluoro-2-methoxy-phenyl boronic acid, 4-fluoro-2-methoxyphenyl-boronic acid, 5-fluoroanisole-2-boronic acid, 4-fluoro-2-methoxy phenyl boronic acid, boronic acid, 4-fluoro-2-methoxyphenyl, pubchem19716, acmc-1c2tm | |
| COC1=CC(F)=CC=C1B(O)O | |
| 169.95 | |
| 169.95 | |
| 4-Fluoro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride) |
| 136°C | |
| C7H8BFO3 | |
| 1 g | |
| ADJBXDCXYMCCAD-UHFFFAOYSA-N | |
| (4-fluoro-2-methoxyphenyl)boronic acid | |
| 2782672 | |
| Crystalline Powder |
Safety and Handling
TSCA : No